missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Vertical Clean Bench
Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.
$23,488.47 - $27,187.03
Specifications
| Product Type | Vertical Clean Bench |
|---|---|
| Coating | Powder |
| Filter Type | HEPA |
| Material | Powder-coated Aluminum and Steel |
| Color | Glacier White, Gray |
| Catalog Number | Mfr. No. | Voltage | Includes | Certifications/Compliance | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Voltage | Includes | Certifications/Compliance | Price | Quantity & Availability | |||
|
10304211
|
Labconco™
3660600 |
115 V | Clean Bench | CAN/CSA C22.2, UL |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
10304212
|
Labconco™
3660601 |
115 V | UV Light and Protection Panel | CAN/CSA C22.2, UL |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
10304213
|
Labconco™
3660603 |
115 V | Airflow Monitor | CAN/CSA C22.2, UL |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
10304214
|
Labconco™
3660604 |
115 V | UV Light, Protection Panel, Airflow Monitor | CAN/CSA C22.2, UL |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
10304215
|
Labconco™
3660620 |
230 V | Clean Bench | CE |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
10304216
|
Labconco™
3660621 |
230 V | UV Light and Protection Panel | CE |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
10304217
|
Labconco™
3660623 |
230 V | Airflow Monitor | CE |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
10304218
|
Labconco™
3660624 |
230 V | UV Light, Protection Panel, Airflow Monitor | CE |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.
- Applications for clean benches include pharmacy compounding, plant tissue culture, media plate preparation, electronics inspection, medical device assembly and more.
- Because they do not provide protection to the user, Vertical Clean Benches should not be used in conjunction with biohazardous material, toxins or radionuclides.
Specifications
| Vertical Clean Bench | |
| HEPA | |
| Glacier White, Gray | |
| 64.3 cm | |
| 30.2 in. | |
| None | |
| 70.8 in. |
| Powder | |
| Powder-coated Aluminum and Steel | |
| 25.3 in. | |
| Vertical Laminar Flow | |
| 81.3 cm | |
| 6 ft. | |
| 179.8 cm |