missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Vertical Clean Bench

Catalog No. 10304218
Change view
Click to view available options
Voltage:
115 V
230 V
Includes:
Airflow Monitor
Clean Bench
UV Light and Protection Panel
UV Light, Protection Panel, Airflow Monitor
Certifications/Compliance:
CAN/CSA C22.2, UL
CE
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage Includes Certifications/Compliance
10304218 230 V UV Light, Protection Panel, Airflow Monitor CE
10304211 115 V Clean Bench CAN/CSA C22.2, UL
10304212 115 V UV Light and Protection Panel CAN/CSA C22.2, UL
10304213 115 V Airflow Monitor CAN/CSA C22.2, UL
10304214 115 V UV Light, Protection Panel, Airflow Monitor CAN/CSA C22.2, UL
10304215 230 V Clean Bench CE
10304216 230 V UV Light and Protection Panel CE
10304217 230 V Airflow Monitor CE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 10304218 Supplier Labconco™ Supplier No. 3660624

missing translation for 'validMainframeMsg'

Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.

  • Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.
  • Applications for clean benches include pharmacy compounding, plant tissue culture, media plate preparation, electronics inspection, medical device assembly and more.
  • Because they do not provide protection to the user, Vertical Clean Benches should not be used in conjunction with biohazardous material, toxins or radionuclides.

Specifications

Product Type Vertical Clean Bench
Coating Powder
Filter Type HEPA
Lighting Type UV
Material Powder-coated Aluminum and Steel
Certifications/Compliance CE
Color Glacier White, Gray
Depth (English) Interior 25.3 in.
Depth (Metric) Interior 64.3 cm
Description Vertical Laminar Flow
Height (English) Interior 30.2 in.
Height (Metric) Interior 81.3 cm
Includes UV Light, Protection Panel, Airflow Monitor
IP Rating None
Voltage 230 V
Width (English) 6 ft.
Width (English) Interior 70.8 in.
Width (Metric) Interior 179.8 cm
Show More Show Less