missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Vertical Clean Bench

Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.

Supplier:  Labconco™ 3660621

 View more versions of this product

Catalog No. 10304216

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.
  • Applications for clean benches include pharmacy compounding, plant tissue culture, media plate preparation, electronics inspection, medical device assembly and more.
  • Because they do not provide protection to the user, Vertical Clean Benches should not be used in conjunction with biohazardous material, toxins or radionuclides.
Specifications

Chemical Identifiers

Vertical Clean Bench
HEPA
Glacier White, Gray
64.3 cm
30.2 in.
None
70.8 in.
Powder
Powder-coated Aluminum and Steel
25.3 in.
Vertical Laminar Flow
81.3 cm
6 ft.
179.8 cm

Specifications

Vertical Clean Bench
HEPA
Powder-coated Aluminum and Steel
Glacier White, Gray
64.3 cm
30.2 in.
UV Light and Protection Panel
230 V
70.8 in.
Powder
UV
CE
25.3 in.
Vertical Laminar Flow
81.3 cm
None
6 ft.
179.8 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions