missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Vertical Clean Bench
Description
- Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.
- Applications for clean benches include pharmacy compounding, plant tissue culture, media plate preparation, electronics inspection, medical device assembly and more.
- Because they do not provide protection to the user, Vertical Clean Benches should not be used in conjunction with biohazardous material, toxins or radionuclides.
Specifications
Specifications
| Product Type | Vertical Clean Bench |
| Coating | Powder |
| Filter Type | HEPA |
| Lighting Type | UV |
| Material | Powder-coated Aluminum and Steel |
| Certifications/Compliance | CAN/CSA C22.2, UL |
| Color | Glacier White, Gray |
| Depth (English) Interior | 25.3 in. |
| Depth (Metric) Interior | 64.3 cm |
| Description | Vertical Laminar Flow |
| Show More |