missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Vertical Clean Bench
Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.
Supplier: Labconco™ 3660620
Description
- Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination.
- Applications for clean benches include pharmacy compounding, plant tissue culture, media plate preparation, electronics inspection, medical device assembly and more.
- Because they do not provide protection to the user, Vertical Clean Benches should not be used in conjunction with biohazardous material, toxins or radionuclides.
Identifiants chimiques
| Vertical Clean Bench | |
| HEPA | |
| Glacier White, Gray | |
| 64.3 cm | |
| 30.2 in. | |
| None | |
| 70.8 in. |
| Powder | |
| Powder-coated Aluminum and Steel | |
| 25.3 in. | |
| Vertical Laminar Flow | |
| 81.3 cm | |
| 6 ft. | |
| 179.8 cm |
Spécifications
| Vertical Clean Bench | |
| HEPA | |
| Powder-coated Aluminum and Steel | |
| Glacier White, Gray | |
| 64.3 cm | |
| 30.2 in. | |
| Clean Bench | |
| 230 V | |
| 70.8 in. |
| Powder | |
| Fluorescent | |
| CE | |
| 25.3 in. | |
| Vertical Laminar Flow | |
| 81.3 cm | |
| None | |
| 6 ft. | |
| 179.8 cm |