missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Zirconium Standard for ICP 1000 ppm 5% HCl/HF, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010331250N
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 92.41%,4.90%,0.49%,0.20% | |
| 1 | |
| Cl2H16O9Zr | |
| UN3264 | |
| Standard | |
| O.Cl[Zr](Cl)=O | |
| 322.24 | |
| 159678 |
| 13520-92-8 , 7647-01-0 , 7664-39-3 , 7732-18-5 | |
| 96.19%,5.10%,0.51%,0.20% | |
| Liquid | |
| MFCD00149898 | |
| Approximately 0°C | |
| SDFOCNSOSARAKC-UHFFFAOYSA-L | |
| oxozirconium;octahydrate;dihydrochloride | |
| 250 mL | |
| High Purity |
Chemical Identifiers
| 13520-92-8 | |
| 322.24 | |
| SDFOCNSOSARAKC-UHFFFAOYSA-L | |
| oxozirconium;octahydrate;dihydrochloride |
| Cl2H16O9Zr | |
| MFCD00149898 | |
| 159678 | |
| O.Cl[Zr](Cl)=O |