missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Zirconium Standard for ICP 1000 ppm 5% HCl/HF, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$458.20 - $1175.67
Chemical Identifiers
| CAS | 13520-92-8 |
|---|---|
| Molecular Formula | Cl2H16O9Zr |
| Molecular Weight (g/mol) | 322.24 |
| MDL Number | MFCD00149898 |
| InChI Key | SDFOCNSOSARAKC-UHFFFAOYSA-L |
| PubChem CID | 159678 |
| IUPAC Name | oxozirconium;octahydrate;dihydrochloride |
| SMILES | O.Cl[Zr](Cl)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010331100
|
Ricca Chemical Company
RV010331100N |
100 mL |
Each for $458.20
|
|
|||||
|
RV010331250
|
Ricca Chemical Company
RV010331250N |
250 mL |
Each for $822.33
|
|
|||||
|
RV010331500
|
Ricca Chemical Company
RV010331500N |
500 mL |
Each for $1,175.67
|
|
|||||
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 13520-92-8 | |
| 322.24 | |
| SDFOCNSOSARAKC-UHFFFAOYSA-L | |
| oxozirconium;octahydrate;dihydrochloride |
| Cl2H16O9Zr | |
| MFCD00149898 | |
| 159678 | |
| O.Cl[Zr](Cl)=O |
Specifications
| Natural LDPE Bottle | |
| 92.41%,4.90%,0.49%,0.20% | |
| 1 | |
| Cl2H16O9Zr | |
| UN3264 | |
| Standard | |
| O.Cl[Zr](Cl)=O | |
| 322.24 | |
| 159678 |
| 13520-92-8 , 7647-01-0 , 7664-39-3 , 7732-18-5 | |
| 96.19%,5.10%,0.51%,0.20% | |
| Liquid | |
| MFCD00149898 | |
| Approximately 0°C | |
| SDFOCNSOSARAKC-UHFFFAOYSA-L | |
| oxozirconium;octahydrate;dihydrochloride | |
| 100 mL | |
| High Purity |