missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Zirconium Standard for ICP 1000 ppm in 2% HNO3/HF, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$458.20 - $1175.67
Chemical Identifiers
| CAS | 14985-18-3 |
|---|---|
| Molecular Formula | N2O7Zr |
| Molecular Weight (g/mol) | 231.23 |
| MDL Number | MFCD00149896 |
| InChI Key | JWBWFTANBHYHBC-UHFFFAOYSA-N |
| PubChem CID | 16211674 |
| IUPAC Name | (nitrooxy)(oxo)zirconio nitrate |
| SMILES | [O-][N+](=O)O[Zr](=O)O[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010362100
|
Ricca Chemical Company
RV010362100N |
100 mL |
Each for $458.20
|
|
|||||
|
RV010362250
|
Ricca Chemical Company
RV010362250N |
250 mL |
Each for $822.33
|
|
|||||
|
RV010362500
|
Ricca Chemical Company
RV010362500N |
500 mL |
Each for $1,175.67
|
|
|||||
Description
- 1000 ppm in 2% HNO3/HF
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 14985-18-3 | |
| 231.23 | |
| JWBWFTANBHYHBC-UHFFFAOYSA-N | |
| (nitrooxy)(oxo)zirconio nitrate |
| N2O7Zr | |
| MFCD00149896 | |
| 16211674 | |
| [O-][N+](=O)O[Zr](=O)O[N+]([O-])=O |
Specifications
| Natural LDPE Bottle | |
| 95.29%,1.96%,0.49%,0.26% | |
| 1 | |
| N2O7Zr | |
| Approximately 0°C | |
| JWBWFTANBHYHBC-UHFFFAOYSA-N | |
| (nitrooxy)(oxo)zirconio nitrate | |
| 100 mL | |
| High Purity |
| 14985-18-3 , 7697-37-2 , 7664-39-3 , 7732-18-5 | |
| 99.17%,2.04%,0.51%,0.28% | |
| Liquid | |
| MFCD00149896 | |
| Standard | |
| [O-][N+](=O)O[Zr](=O)O[N+]([O-])=O | |
| 231.23 | |
| 16211674 |