Learn More
United Scientific™ Variable Volume, Fully Autoclavable Micropipettes
These Variable Volume, Fully Autoclavable Micropipettes are highly accurate and easy to calibrate/maintain.
Supplier:  United Scientific™ PRV1000
 Afficher plus de versions de ce produit
Numéro de catalogue. S43752
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Built-in tip ejectors and autoclavable tip cones work on a click stop digital system.
  • Easy to disassemble prior to autoclaving.
  • Manufactured in an ISO 9001:2008 certified facility.
  • Pipettes are individually calibrated according to ISO 8655 standards.
  • A calibration certificate is provided with each pipette.
Spécifications
Spécifications
Autoclavable
Calibrated According to ISO 8655 Standards
Non-sterile
No
Digital
Blue
228 mm
Reusable
Yes
Plastic
For use with 1000 μL Tips
10 μL
Calibration Certificate
100 to 1000 μL
Volumetric Pipet
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS