missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Vanadium(III) chloride DFS Item

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

$494.30 - $494.30

Spécifications

CAS 7718-98-1
Molecular Formula Cl3V
Linear Formula VCl3
MDL Number MFCD00011454
Synonym Vanadium trichloride
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
208272100G DFS Item
Afficher les documents
Sigma Aldrich
208272100G
100 g
chaque for $494.30
Il en reste null
 
Description

Description

Vanadium trichloride acts as a reducing agent.Vanadium chloride may be used as a reduction agent to determine nitrates and nitrites in water samples.It may be used in redox flow batteries. Vanadium trichloride catalyses Pinacol coupling reactions of benzaldehyde in water. it also acts as a mild and efficient catalyst for the chemoselective deprotection of acetonides.
Spécifications

Spécifications

7718-98-1
VCl3
Vanadium trichloride
97%
Cl3V
MFCD00011454
157.3
3 g/mL (at 25°C)
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'