Learn More
Valeramide, 97%
CAS: 626-97-1 | C5H11NO | 101.15 g/mol
$443.67 - $443.67
Identifiants chimiques
| CAS | 626-97-1 |
|---|---|
| Molecular Formula | C5H11NO |
| Molecular Weight (g/mol) | 101.15 |
| MDL Number | MFCD00041895 |
| InChI Key | IPWFJLQDVFKJDU-UHFFFAOYSA-N |
| Synonym | valeramide, n-valeramide, pentanimidic acid, unii-tkl9tn43qk, tkl9tn43qk, ksc353i5p, ipwfjlqdvfkjdu-uhfffaoysa, valeramide, pharmaceutical impurity standard |
| PubChem CID | 12298 |
| ChEBI | CHEBI:16459 |
| IUPAC Name | pentanamide |
| SMILES | CCCCC(=O)N |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC422440250
|
Thermo Scientific Chemicals
422440250 |
25 g | Glass bottle |
chaque for $443.67
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 626-97-1 | |
| 101.15 | |
| IPWFJLQDVFKJDU-UHFFFAOYSA-N | |
| 12298 | |
| pentanamide |
| C5H11NO | |
| MFCD00041895 | |
| valeramide, n-valeramide, pentanimidic acid, unii-tkl9tn43qk, tkl9tn43qk, ksc353i5p, ipwfjlqdvfkjdu-uhfffaoysa, valeramide, pharmaceutical impurity standard | |
| CHEBI:16459 | |
| CCCCC(=O)N |
Specifications
| 626-97-1 | |
| White | |
| 96% min. (GC) | |
| C5H11NO | |
| MFCD00041895 | |
| valeramide, n-valeramide, pentanimidic acid, unii-tkl9tn43qk, tkl9tn43qk, ksc353i5p, ipwfjlqdvfkjdu-uhfffaoysa, valeramide, pharmaceutical impurity standard | |
| CCCCC(=O)N | |
| 101.15 | |
| CHEBI:16459 | |
| 97% | |
| Valeramide, 97% |
| 101°C to 106°C | |
| Authentic | |
| Glass bottle | |
| CH3(CH2)3CONH2 | |
| 25 g | |
| IPWFJLQDVFKJDU-UHFFFAOYSA-N | |
| pentanamide | |
| 12298 | |
| 101.15 | |
| Shiny Powder or Flakes |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 210-974-0
TSCA : TSCA
RUO – Research Use Only