missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 5000 Hooded Coverall
Great tear resistance
Supplier: DuPont™ C3127TTNLG000600
Description
- Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
- Attached hood with elastic around face opening.
- Strong zipper for tough, flexible closure
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Manufactured under specifications that do not contain natural rubber latex
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Specifications
| Large | |
| Tychem | |
| Elastic | |
| Hooded | |
| Zipper | |
| Standard fit hood, elastic wrist and ankle; Multilayer composite barrier film laminated to a durable Polypropylene substrate | |
| 94.61 to 103.5 cm | |
| 34 in. | |
| 25.25 in. | |
| 1.67 to 1.8 m |
| Unisex | |
| Tan | |
| Elastic | |
| Standard | |
| Taped | |
| 29 in. | |
| Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness | |
| 86.36 cm | |
| 5.5 to 5.9 ft. |