missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tween 20 washing buffer, 0.5% in PBS (10X)

$207.60 - $382.99

Spécifications

Chemical Name or Material Blocking Buffer
Concentration 0.5% in PBS (10X)
Physical Form Liquid
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
AAJ60304K2
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
J60304K2
1 L
chaque for $207.60
Il en reste null
 
AAJ60304K3
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
J60304K3
2 L
chaque for $382.99
Il en reste null
 
Description

Description

Tween 20 blocking buffer solution is used as a washing agent in immunoassays, such as Western blots and ELISAs. It helps to prevent non-specific antibody binding. The buffer is used for washes between each immunoreactions, to remove unbound immunologicals, and eventually for incubation solutions of immunoreagents (labeled antibodies) to reduce unspecific background.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Blocking Buffer
0.5% in PBS (10X)
Liquid
Videos
Sécurité et manipulation

Sécurité et manipulation

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

RUO – Research Use Only