Learn More
Thermo Scientific™ Tryptic Soy Broth
Cultivate and isolate microorganisms with ease using this general purpose enrichment broth with formats available that meet USP guidelines for sterility testing.
Supplier:  Thermo Scientific™ R07228
 Afficher plus de versions de ce produit
Numéro de catalogue. R07228
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Cultivate and isolate microorganisms with ease using Thermo Scientific™ Tryptic Soy Broth, a general purpose enrichment broth.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use TSB tubes.
Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Spécifications
Spécifications
AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP
2 to 25°C
Tube
Microbiology Media
Glass tube with black screw cap
Clear liquid, light to medium amber
Tryptic Soy Broth
United States
10 mL
20/Pk.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS