missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethoxyethylsilane 95.0+%, TCI America™
Supplier: TCI America E018925ML
Spécifications
| Triethoxyethylsilane | |
| -78°C | |
| 161°C | |
| MFCD00026754 | |
| 1993 | |
| DENFJSAFJTVPJR-UHFFFAOYSA-N | |
| triethoxy(ethyl)silane | |
| 6515 | |
| ≥95.0% (GC) |
| 78-07-9 | |
| Colorless | |
| C8H20O3Si | |
| 25 mL | |
| ethyltriethoxysilane, triethoxy ethyl silane, silane, triethoxyethyl, a 15 silane, union carbide a-15, triethoxy-ethylsilane, silane, ethyltriethoxy, silane, ethyl triethoxy a-15, ethaneorthosiliconic acid, triethyl ester, ethyl triethoxysilane | |
| CCO[Si](CC)(OCC)OCC | |
| 192.33 | |
| 192.33 | |
| Liquid |
Identifiants chimiques
| 78-07-9 | |
| 192.33 | |
| DENFJSAFJTVPJR-UHFFFAOYSA-N | |
| 6515 | |
| CCO[Si](CC)(OCC)OCC |
| C8H20O3Si | |
| MFCD00026754 | |
| ethyltriethoxysilane, triethoxy ethyl silane, silane, triethoxyethyl, a 15 silane, union carbide a-15, triethoxy-ethylsilane, silane, ethyltriethoxy, silane, ethyl triethoxy a-15, ethaneorthosiliconic acid, triethyl ester, ethyl triethoxysilane | |
| triethoxy(ethyl)silane |
Sécurité et manipulation
missing translation for 'einecsNumber' : (2)-2052&(2)-2053
missing translation for 'rtecsNumber' : VV4205000
missing translation for 'tsca' : Yes