missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tricine, 0.5M buffer soln., pH 8.0
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Color | Colorless |
| Concentration | 0.5 M |
| pH | 8.0 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Chemical Name or Material | Tricine buffer soln. |
Sécurité et manipulation
| Hazard Category | H335 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only