Learn More
trans-dia.mminedibromopalladium(II), 99.99% (metals basis), Pd 34.7% min, Thermo Scientific™, Premion™
Supplier: Thermo Scientific Chemicals 3924903
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
| trans-Diamminedibromopalladium(II) | |
| 290°C (decomposition) | |
| 99.99% (metals basis) | |
| MFCD00058860 | |
| dibromopalladium diamine, trans-diamminedibromopalladium ii , premion, palladium,diamminedibromo-, sp-4-1-9ci | |
| N.N.[Br-].[Br-].[Pd++] | |
| 300.29 | |
| 300.28 |
| 14591-90-3 | |
| 2.5 g/mL | |
| Br2H6N2Pd | |
| 1 g | |
| WLAAECNKUYLOJV-UHFFFAOYSA-L | |
| dibromopalladium diamine | |
| 15336283 | |
| Powder |
Identifiants chimiques
| 14591-90-3 | |
| 300.29 | |
| WLAAECNKUYLOJV-UHFFFAOYSA-L | |
| 15336283 | |
| N.N.[Br-].[Br-].[Pd++] |
| Br2H6N2Pd | |
| MFCD00058860 | |
| dibromopalladium diamine, trans-diamminedibromopalladium ii , premion, palladium,diamminedibromo-, sp-4-1-9ci | |
| dibromopalladium diamine |
Sécurité et manipulation
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures