missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-Ethylcyclohexanecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America E09045G
Spécifications
| trans-4-Ethylcyclohexanecarboxylic Acid | |
| 52°C | |
| C9H16O2 | |
| 5 g | |
| UNROFSAOTBVBBT-UHFFFAOYSA-N | |
| 4-ethylcyclohexane-1-carboxylic acid | |
| 1490288 | |
| ≥98.0% (GC,T) |
| 6833-47-2 | |
| White | |
| MFCD01076323,MFCD01568857 | |
| trans-4-ethylcyclohexanecarboxylic acid, 4-ethylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-ethyl, 4-trans-ethyl cyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-ethyl-, trans, 1s,4r-4-ethylcyclohexane-1-carboxylic acid, pubchem2491, acmc-209o2z, ksc352s1t, ksc352s2b | |
| CCC1CCC(CC1)C(O)=O | |
| 156.23 | |
| 156.23 | |
| Crystalline Powder |
Identifiants chimiques
| 6833-47-2 | |
| 156.23 | |
| UNROFSAOTBVBBT-UHFFFAOYSA-N | |
| 1490288 | |
| CCC1CCC(CC1)C(O)=O |
| C9H16O2 | |
| MFCD01076323,MFCD01568857 | |
| trans-4-ethylcyclohexanecarboxylic acid, 4-ethylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-ethyl, 4-trans-ethyl cyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-ethyl-, trans, 1s,4r-4-ethylcyclohexane-1-carboxylic acid, pubchem2491, acmc-209o2z, ksc352s1t, ksc352s2b | |
| 4-ethylcyclohexane-1-carboxylic acid |
Sécurité et manipulation
missing translation for 'tsca' : No