missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Data Logging Ethernet Thermometers

Fisherbrand™ Traceable™ Data Logging Ethernet Thermometers with TraceableLive™ Cloud Service compatibility

$972.82 - $1,446.27

Spécifications

Type Temperature Datalogger
Display Type LCD
Calibration Calibrated
Alarm Types Audible, Visual
Software Type TraceableLIVE
View More Specs

Products 2
missing translation for 'productCode' missing translation for 'brand' Includes Price Quantity & Availability  
missing translation for 'productCode' missing translation for 'brand' Includes Price Quantity & Availability  
15079704
missing translation for 'viewDocuments' Promotion Details
Fisherbrand™
6101217
Bottle Probe with 10 ft. cable (1), Magnetic Back Holder, Ethernet Cable, Universal Power Adapter with 5 ft. cord, AAA alkaline batteries (3)
chaque for $972.82
Only null left
 
15079705
missing translation for 'viewDocuments' Promotion Details
Fisherbrand™
6101211
Stainless Steel Probe with 10 ft. cable (1), Magnetic Back Holder, Ethernet Cable, Universal Power Adapter with 5 ft. cord, AAA alkaline batteries (3)
chaque for $1,446.27
Only null left
 
Description

Description

  • Connect to your data via smartphone, tablet, or PC using TraceableLive™ Cloud Service
  • Get the reliability, speed, and security using Ethernet connection
  • View current temperature conditions anywhere in the world
  • Cloud-based interface sends email, text, and push notifications
  • No software required: Web, Android™, Apple™ iOS data visibility
  • Get accurate, reliable measurements with included NIST-traceable calibration
Specifications

Specifications

Temperature Datalogger
Calibrated
TraceableLIVE
Plastic
±0.10°C
6.985 cm
LCD
Audible, Visual
NIST-Traceable Calibration
Temperature Monitoring
0.01°C
0.45 kg
Videos
SDS
Documents

Documents