missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.98% (metals basis), Thermo Scientific Chemicals
$162.05 - $162.05
Identifiants chimiques
| CAS | 7440-32-6 |
|---|---|
| Molecular Formula | Ti |
| Molecular Weight (g/mol) | 47.87 |
| MDL Number | MFCD00011264,MFCD00151301 |
| InChI Key | RTAQQCXQSZGOHL-UHFFFAOYSA-N |
| Synonym | oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 |
| PubChem CID | 23963 |
| ChEBI | CHEBI:33341 |
| IUPAC Name | titanium |
| SMILES | [Ti] |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AA42388QK
|
Thermo Scientific Chemicals
042388QK |
25 Slugs |
chaque for $162.05
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 7440-32-6 | |
| 47.87 | |
| RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
| 23963 | |
| titanium |
| Ti | |
| MFCD00011264,MFCD00151301 | |
| oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
| CHEBI:33341 | |
| [Ti] |
Spécifications
| 7440-32-6 | |
| Odorless | |
| Slug | |
| MFCD00011264,MFCD00151301 | |
| RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
| titanium | |
| 23963 | |
| 99.98% | |
| (metals basis) |
| 25 Slugs | |
| ≈0.9g/slug | |
| Ti | |
| oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
| [Ti] | |
| 47.87 | |
| CHEBI:33341 | |
| Titanium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length |
Sécurité et manipulation
missing translation for 'einecsNumber' : 231-142-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only