Learn More
Honeywell™ Titan™ Non-Stretch Harnesses
Constructed of tough polyester webbing
Supplier: Honeywell™ T4007UAKC
Comprend:
Trilingual labeling and instructions.
Description
Fully adjustable harnesses feature mating buckles on the chest and shoulder straps, and a sliding back D-ring with a flexible D-ring pad.
- Sub-pelvic strap reduces pressure on shoulders
- Available with mating or tongue buckle leg straps and optional side D-rings
Order Info
Universal size fits most.
Compliance
Harnesses meet applicable OSHA, ANSI™, and CSA standards.
Spécifications
| Mating | |
| Universal | |
| Sliding Back, Side |
| Orange | |
| Polyester Webbing | |
| Meets applicable OSHA, ANSI, and CSA standards. |