missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCI America™ Tissue-Clearing Reagent CUBIC-B
CUBIC-B can be used for clearing bones as an optional step used alongside the regular CUBIC protocol.
$102.32 - $336.17
Spécifications
| For Use With (Equipment) | Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device |
|---|---|
| Product Line | CUBIC |
| For Use With (Application) | Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications |
| Form | Liquid |
| Numéro de catalogue | Numéro du manufacturier. | Capacity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Capacity | Prix | Quantité | |||||
|
T378025ML
|
TCI America™
T378025ML |
25 mL |
chaque for $102.32
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
T3780100ML
|
TCI America™
T3780100ML |
100 mL |
chaque for $336.17
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
- CUBIC-B is an optional addition to the CUBIC clearing protocol, and can be used to clear bone tissue.
Note: Both T3741 and T3740 are required for the CUBIC clearing process.
Spécifications
| Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device | |
| CUBIC | |
| Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications | |
| Liquid |