missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TISAB II, For Fluoride, Certified, LabChem™
Supplier: LabChem LC261305
Specifications
| TISAB II | |
| For Fluoride | |
| 7647-14-5 , 64-19-7 , 1310-73-2 , 125572-95-4 | |
| Soluble in water | |
| Certified | |
| Cubitainer™ | |
| Carbon monoxide; Carbon dioxide | |
| 20 L |
| Colorless | |
| 83.7,5.8,5.7,4.4,0.4 | |
| CH3COOH | |
| 60.05 | |
| Passes Test | |
| Traceable to NIST | |
| 5.3 to 5.5 | |
| Liquid |