Learn More
Labcon North America ZAP™ 10uL Aerosol Filter Pipet Tips, in Racks, Sterile DFS Item
Supplier:  Labcon North America 10519650389
Tip, Pipet; Labcon; ZAP™ Aerosol Filter; Fits most popular 10uL Pipettor; Sterile; Patented robotically inserted endotoxin free filter; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Refillable autoclavable racks; 96 tips/rack; 12 racks/pack; 10 packs/case; 31.4mm H.; Made in USA
Numéro de catalogue. 50101574
Il en reste null
Découvrez plus d'offres spéciales