missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Hydroxide (10% in Water) [Reagent for Ion-Pair Chromatography], TCI America™

Supplier:  TCI America I036325ML

Numéro de catalogue. I036325ML


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Tetraethylammonium Hydroxide (10% in Water) [Reagent for Ion-Pair Chromatography]
Colorless
MFCD00009024
3267
LRGJRHZIDJQFCL-UHFFFAOYSA-M
tetraethylazanium hydroxide
6509
Liquid
77-98-5
C8H21NO
25 mL
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
[OH-].CC[N+](CC)(CC)CC
147.26
147.26

Identifiants chimiques

77-98-5
147.26
LRGJRHZIDJQFCL-UHFFFAOYSA-M
6509
[OH-].CC[N+](CC)(CC)CC
C8H21NO
MFCD00009024
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
tetraethylazanium hydroxide
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : (2)-0186

missing translation for 'rtecsNumber' : KH3150000

missing translation for 'tsca' : Yes

FDS
Documents

Documents

Promotions

Promotions