Learn More
Fisherbrand™ Test Paper
Use these test papers for a variety of applications.
$42.85 - $68.38
Produits 5
Numéro de catalogue Numéro du manufacturier. Type Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Type Prix Quantité  
S60100
Afficher les documents
Fisherbrand™
15012V100
Cobalt Chloride Test Paper
paquet de 12 chaque for $68.38
Il en reste null
S67172
Afficher les documents
Fisherbrand™
18512V100
Litmus Neutral Test Strip
paquet de 12 chaque for $59.36
Il en reste null
S60130
Afficher les documents
Fisherbrand™
16512V100
PTC Taste Test Paper
paquet de 12 chaque for $68.38
Il en reste null
S67171
Afficher les documents
Fisherbrand™
19012V100
Litmus Red Test Strip
paquet de 12 chaque for $42.85
Il en reste null
S60126
Afficher les documents
Fisherbrand™
13512V100
Moisture Detection Test Strip
paquet de 12 chaque for $68.38
Il en reste null
Description
Description
  • PTC (pheylthiocarbamide) taste test paper test for the genetically controlled ability to taste this substance
  • This will allow for the development of a Pedigree for an individual
  • PTC can taste bland, bitter or even vile depending upon how sensitive your taste buds are to this chemical
  • The bitter taste detected by some individuals, and its intensity, depends upon a homozygous dominant or heterozygous condition in the individuals tested
  • PTC Taste Test Paper is perfectly safe to use
  • Phenylthiocarbamide (PTC) is present at only 3-5 micrograms per strip
  • At this level, the compound is negligible and harmless
  • Cobalt Chloride test paper detects the presence of moisture
  • This test paper is useful in difficult to reach places and recesses
  • Cobalt chloride test paper is responsive to 50% relative humidity and above, and any direct measure of an aqueous solution
  • Phenolphthalein test paper is sensitive to Ammonia Vapors, and thus is used to detect Ammonia leaks, for example, in refrigerators or air conditioners
  • Phenolphthalein detects Ammonia in solution from 3 - 5ppm
  • The test strip will turn a pinkish color if Ammonia is detected
Videos
FDS