missing translation for 'onlineSavingsMsg'
Learn More

TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™

Numéro de catalogue. p-30476
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
BP2471100 100 mL Poly Bottle
BP2471500 500 mL Poly Bottle
BP24711 1 L Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. BP2471100 Fournisseur Fisher BioReagents Code fournisseur BP2471100
Il en reste null

TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology

  • Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X TBS buffer solution - simply dilute as needed
  • 10X solution contains 1.37M NaCl, 0.027M KCl and 0.25M tris
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TBS solution with 9 volumes of ultrapure water, such as BP2819
  • Filtered and autoclaved - ready to use stock TBS buffer
  • Tested for the absence of DNase, RNase, and protease
  • TBS buffer is often used in immunodetection techniques such as Western blotting protocols
  • Intended for laboratory research use only

Spécifications

Chemical Name or Material Tris Buffered Saline
Name Note 10X Solution
CAS 7447-40-7, 7647-01-0, 7647-14-5, 77-86-1, 7732-18-5
Boiling Point 100°C
Color Colorless
pH 7.5
Physical Form Liquid
Quantity 100 mL
DNase DNase free
Purity Grade Notes DNase-, RNase- and Protease-Free
Packaging Poly Bottle
Protease Protease free
Synonym TBS
Grade Molecular Biology
Concentration or Composition (by Analyte or Components) 10X solution contains 1.37M Sodium Chloride, 0.027M Potassium Chloride, and 0.25M Tris/Tris-HCl.
Afficher plus de résultats Afficher moins de résultats