missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) oxide, 99.99%, (trace metal basis)

CAS: 1314-61-0 | O5Ta2 | 441.89 g/mol

$152.92 - $444.62

Identifiants chimiques

CAS 1314-61-0
Molecular Formula O5Ta2
Molecular Weight (g/mol) 441.89
MDL Number MFCD00011254
InChI Key PBCFLUZVCVVTBY-UHFFFAOYSA-N
IUPAC Name [(dioxotantalio)oxy]dioxotantalum
SMILES O=[Ta](=O)O[Ta](=O)=O
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC194170100
FDS Afficher les documents
Thermo Scientific Chemicals
194170100
10 g Glass bottle
chaque for $152.92
Il en reste null
 
AC194170500
FDS Afficher les documents
Thermo Scientific Chemicals
194170500
50 g Glass bottle
chaque for $444.62
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Spécifications

Identifiants chimiques

1314-61-0
441.89
PBCFLUZVCVVTBY-UHFFFAOYSA-N
O=[Ta](=O)O[Ta](=O)=O
O5Ta2
MFCD00011254
[(dioxotantalio)oxy]dioxotantalum

Spécifications

1872°C
1314-61-0
10 g
O5Ta2
MFCD00011254
Solubility in water: insoluble
O=[Ta](=O)O[Ta](=O)=O
441.89
99.99%
Glass bottle
Beige-White to White
Powder
99.99%
Ta2O5
15, 9188
PBCFLUZVCVVTBY-UHFFFAOYSA-N
[(dioxotantalio)oxy]dioxotantalum
441.89
Trace Metal Basis
Tantalum(V) oxide
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

missing translation for 'einecsNumber' : 215-238-2

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only