Learn More
Chemglass Life Sciences Syringe, Luer Lock, 50cc Capacity, 2cc Subdivisions DFS Item
Supplier:  Chemglass Inc CG307007
Syringe, Luer Lock, 50cc Capacity, 2cc Subdivisions. Borosilicate glass syringe has interchangeable barrels and plungers. Colored plunger ends identify the capacity of the syringe. Flanges are flat on two sides to prevent syringe from rolling. All sizes of syringes are equipped with a metal luer lock and permanently fused graduations. For needles see CG-3075.
Numéro de catalogue. CG307007
Il en reste null
Découvrez plus d'offres spéciales