Learn More
Synthware™ Single Neck Round Bottom Flasks, Full Length Joint
Hand-blown from tubing to ensure uniform wall thickness
Supplier:  Synthware™ F30291L
 View more versions of this product
Catalog No. 31501127
Only null left
Explore available promotions
Description
Description
SYNTHWARE round bottom boiling flasks with an extended neck and full length standard taper outer joint. All sizes are heavy wall design and are hand blown from borosilicate tubing to ensure uniform wall thickness.
Specifications
Specifications
Clear
1000 mL
Organic Synthesis
Single neck, round bottom flask
Borosilicate Glass (3.3)
29/42
Product Suggestions
Product Suggestions