missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Single Neck Round Bottom Flasks, Full Length Joint

Description
SYNTHWARE round bottom boiling flasks with an extended neck and full length standard taper outer joint. All sizes are heavy wall design and are hand blown from borosilicate tubing to ensure uniform wall thickness.
Specifications
Specifications
| Color | Clear |
| Description | Single neck, round bottom flask |
| Capacity (Metric) | 25 mL |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Organic Synthesis |
| Standard Taper Joint | 24/40 |