Learn More
Synthware™ Single Neck Round Bottom Flasks, Full Length Joint
Hand-blown from tubing to ensure uniform wall thickness
Supplier:  Synthware™ F30242L
 Afficher plus de versions de ce produit
Numéro de catalogue. 31501128
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
SYNTHWARE round bottom boiling flasks with an extended neck and full length standard taper outer joint. All sizes are heavy wall design and are hand blown from borosilicate tubing to ensure uniform wall thickness.
Spécifications
Spécifications
Clear
2000 mL
Organic Synthesis
Single neck, round bottom flask
Borosilicate Glass (3.3)
24/40
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS