Learn More
Synthware™ Chromatography Column with Spherical Joint and PTFE stopcock
General purpose chromatography column for flash chromatography
Supplier: Synthware™ C195324
Description
- Equipped with PTFE stopcock and top 35/20 socket joint
- Synthware columns designed with heavy-walls for increased safety
Spécifications
| 26 mm | |
| 35/20 | |
| 457 mm |
| Spherical joint | |
| 2 mm bore |