missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ IV One-Step RT-PCR System

Numéro de catalogue. p-8385018
Encompass
Modifier l'affichage
Click to view available options
Includes:
Kit only
Kit with ezDNase
No. of Reactions:
100 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Includes No. of Reactions
12594100 Kit only 100 Reactions
12595100 Kit with ezDNase 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 12594100 Fournisseur Invitrogen™ Code fournisseur 12594100
Il en reste null

Comprend

  • SuperScript IV RT Mix, 12.5µL
  • 2X Platinum SuperFi RT-PCR Master Mix, 625µL
  • Nuclease-free water, 1.25mL

The Invitrogen SuperScript One-Step RT-PCR System, available with or without ezDNase, combines the high processivity of SuperScript IV Reverse Transcriptase (RT) with the high-fidelity of Platinum SuperFi DNA Polymerase to provide unmatched one-step RT-PCR performance.

The Invitrogen SuperScript One-Step RT-PCR System, available with or without ezDNase, combines the high processivity of SuperScript IV Reverse Transcriptase (RT) with the high-fidelity of Platinum SuperFi DNA Polymerase to provide unmatched one-step RT-PCR performance. With this convenient one-step formulation plus gene-specific primers, you can perform both cDNA synthesis from total RNA or mRNA and PCR amplification in a single tube.

Features of the SuperScript IV One-Step RT-PCR System include:

  • Superior performance with sensitivity down to 0.01 pg RNA, target length up to 13.8 kb, and the fastest one-step RT-PCR protocol
  • Reliable target detection even with suboptimally pure RNA samples
  • Two-phase hot-start activation mechanism for high sensitivity, specificity, and room temperature setup
  • Fast and easy gDNA removal for superior accuracy and confidence in results (SuperScript IV One-Step RT-PCR System with ezDNase only)

SuperScript IV RT is a proprietary MMLV RT mutant with increased processivity, thermostability, and inhibitor resistance, as well as reduced RNase H activity, for sensitive and efficient full-length cDNA synthesis. Platinum SuperFi DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with trusted Platinum hot-start technology for the highest success in PCR. The combination of these two engineered enzymes produces enhanced RT-PCR yields and fidelity, as well as amplification of longer templates compared to other one-step RT-PCR kits.

The inclusion of ezDNase (SuperScript IV One-Step RT-PCR System with ezDNase only) for use upstream of the RT step further enhances the one-step RT-PCR workflow through an extremely simplified genomic DNA removal step that ensures the highest accuracy and confidence in one-step RT-PCR results.

Spécifications

Content And Storage

• SuperScript IV RT Mix (50 μL)
• 2X Platinum SuperFi RT-PCR Master Mix (2 x 1.25 mL)
• Nuclease-free water (2 x 1.25 mL)

Store at –20°C.

Detection Method Gel Electrophoresis
Format Kit
GC-Rich PCR Performance High
PCR Method 1-step RT-PCR
Polymerase Platinum SuperFi
Reaction Speed 10 min.
Technique 1-step RT-PCR
Optimal Reaction Temperature 45°C to 60°C
Quantity 1 Ea.
Reverse Transcriptase SuperScript IV
Ribonuclease H Activity Reduced
Shipping Condition Dry Ice
Final Product Type PCR Amplified cDNA
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Reaction Format Master Mix
Reagent Type Reverse Transcription
Size (Final Product) Up to 14 kb
Starting Material RNA
Includes Kit only
Afficher plus de résultats Afficher moins de résultats

For Research Use Only. Not for use in diagnostic procedures.