missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid solution, 0.1M, Reag. Ph. Eur., Honeywell Fluka™

Volumetric, Reag. Ph. Eur., 0.1 M H2SO4 (0.2N)

$209.13 - $1090.85

Identifiants chimiques

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
6001605
Afficher les documents
Honeywell Inc
353571L
1 L
chaque for $209.13
Il en reste null
 
60046577
Afficher les documents
Honeywell Inc
353576X1L
6 x 1 L
caisse de 6 chaque for $1,090.85
Il en reste null
 
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Spécifications

Identifiants chimiques

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O

Spécifications

7664-93-9
1 L
H2SO4
UN3264
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
Reagent Ph. Eur.
0.1M
H2O4S
MFCD00064589
2037554
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08g/mol
Sulfuric acid solution
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'dotInformation' : 8 / PGIII

FDS
missing translation for 'documents'

missing translation for 'documents'