missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan Red 7B

CAS: 6368-72-5 | C24H21N5 | 379.467 g/mol

$66.73 - $66.73

Identifiants chimiques

CAS 6368-72-5
Molecular Formula C24H21N5
Molecular Weight (g/mol) 379.467
MDL Number MFCD00003904
InChI Key VKWNTWQXVLKCSG-UHFFFAOYSA-N
Synonym C.I. 26050, N-Ethyl-1-[4-(phenylazo)phenylazo]-2-naphthylamine, Fat Red 7B, Solvent Red 19
PubChem CID 61396
IUPAC Name N-ethyl-1-[(4-phenyldiazenylphenyl)diazenyl]naphthalen-2-amine
SMILES CCNC1=C(C2=CC=CC=C2C=C1)N=NC3=CC=C(C=C3)N=NC4=CC=CC=C4
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC191630050
FDS Afficher les documents
Thermo Scientific Chemicals
191630050
5 g Glass bottle
chaque for $66.73
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

6368-72-5
379.467
VKWNTWQXVLKCSG-UHFFFAOYSA-N
61396
CCNC1=C(C2=CC=CC=C2C=C1)N=NC3=CC=C(C=C3)N=NC4=CC=CC=C4
C24H21N5
MFCD00003904
C.I. 26050, N-Ethyl-1-[4-(phenylazo)phenylazo]-2-naphthylamine, Fat Red 7B, Solvent Red 19
N-ethyl-1-[(4-phenyldiazenylphenyl)diazenyl]naphthalen-2-amine

Spécifications

130.0°C
C24H21N5
16, 379
VKWNTWQXVLKCSG-UHFFFAOYSA-N
CCNC1=C(C2=CC=CC=C2C=C1)N=NC3=CC=C(C=C3)N=NC4=CC=CC=C4
379.467
379.45
Complies
5 g
Sudan red 7B
6368-72-5
MFCD00003904
C.I. 26050, N-Ethyl-1-[4-(phenylazo)phenylazo]-2-naphthylamine, Fat Red 7B, Solvent Red 19
Authentic
N-ethyl-1-[(4-phenyldiazenylphenyl)diazenyl]naphthalen-2-amine
61396
Glass bottle
Bordeaux
Crystalline Powder
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

missing translation for 'einecsNumber' : 228-862-5

missing translation for 'rtecsNumber' : QM2850000

missing translation for 'tsca' : TSCA

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only