missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium ICP Standard, 10,000 ppm Sr in 5% HNO3, Ricca Chemical
$1131.77 - $1131.77
Chemical Identifiers
| CAS | 10042-76-9 |
|---|---|
| Molecular Formula | N2O6Sr |
| Molecular Weight (g/mol) | 211.63 |
| MDL Number | MFCD00011248 |
| InChI Key | DHEQXMRUPNDRPG-UHFFFAOYSA-N |
| PubChem CID | 24848 |
| IUPAC Name | strontium(2+) dinitrate |
| SMILES | [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PSR10KN-500
|
Ricca Chemical Company
PSR10KN500 |
500 mL | Natural Poly Bottle |
Each for $1,131.77
|
|
||||
Chemical Identifiers
| 10042-76-9 | |
| 211.63 | |
| DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
| strontium(2+) dinitrate |
| N2O6Sr | |
| MFCD00011248 | |
| 24848 | |
| [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
| Natural Poly Bottle | |
| 10,000ppm Sr | |
| 93.0,6.97,2.24 | |
| Liquid | |
| MFCD00011248 | |
| < 2 | |
| DHEQXMRUPNDRPG-UHFFFAOYSA-N | |
| strontium(2+) dinitrate | |
| 500 mL | |
| Strontium ICP Standard |
| 10042-76-9 , 7697-37-2 , 7732-18-5 | |
| 89.0,6.7,2.15 | |
| 1 mL = 10 mg Sr | |
| N2O6Sr | |
| Colorless | |
| Standard | |
| [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 211.63 | |
| 24848 |