Learn More
Stemcell Technologies STEMdiff™ Cardiomyocyte Dissociation Kit, Size: 1 Kit DFS Item
Supplier:  Stemcell Technologies Inc STM05025
STEMdiff™ Cardiomyocyte Dissociation Kit includes STEMdiff™ Cardiomyocyte Dissociation Medium and STEMdiff™ Cardiomyocyte Support Medium. STEMdiff™ Cardiomyocyte Dissociation Medium can be used to harvest cardiomyocytes that have been differentiated from human pluripotent stem cells (hPSCs) using STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit (Catalog #05010) or STEMdiff™ Atrial Cardiomyocyte Differentiation Kit (Catalog #100-0215) and maintained in STEMdiff™ Cardiomyocyte Maintenance Medium (Catalog #05010/05020). STEMdiff™ Cardiomyocyte Support Medium reduces stress on these cardiomyocytes during harvesting and replating, maintaining their viability and functional capacity for downstream applications and analyses.
Numéro de catalogue. NC1355349
Il en reste null
Découvrez plus d'offres spéciales