Learn More
Thermo Scientific™ StartingBlock™ Blocking Buffer
StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays.
Supplier:  Thermo Scientific™ 37538X3
 Afficher plus de versions de ce produit
Numéro de catalogue. FER37538X3
Il en reste null
Description
Description
StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays. It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock Blocking Buffer is available in PBS and TBS, with and without Tween 20.
Features of StartingBlock Blocking Buffer include:
  • Strip and re-probe without re-blocking—blots stay blocked even after stripping with Restore Stripping Buffer (Cat. No. 21059)
  • Compatible with many detection systems—western blot, ELISA, and IHC with antibody or avidin/biotin probes (blocker is serum- and biotin-free)
  • Short blocking times—less than 15 minutes for nitrocellulose or PVDF membranes; almost instantaneous for polystyrene microplate wells
Spécifications
Identifiants chimiques
Western Blot
Liquid
StartingBlock
Spécifications
PBS
Liquid
3 x 1 L
Western Blot
StartingBlock
Proprietary protein formulation in phosphate-buffered saline, pH 7.5
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
Sécurité et manipulation
Sécurité et manipulation
Recommended Storage : Upon receipt store at 4°C.
FDS