missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ StartingBlock™ Blocking Buffer

Numéro de catalogue. p-7184457
Modifier l'affichage
Click to view available options
Quantity:
100 mL
1 L
3 x 1 L
Chemical Name or Material:
PBS
PBST
TBS
TBST
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Chemical Name or Material
PI37578 100 mL PBS
PI37538 1 L PBS
PI37579 100 mL TBS
FER37538X3 3 x 1 L PBS
PI37542 1 L TBS
EN37539 1 L PBST
EN37543 1 L TBST
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Numéro de catalogue. PI37578 Fournisseur Thermo Scientific™ Code fournisseur 37578
Il en reste null

StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays.

StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays. It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock Blocking Buffer is available in PBS and TBS, with and without Tween 20.

Features of StartingBlock Blocking Buffer include:

  • Strip and re-probe without re-blocking—blots stay blocked even after stripping with Restore Stripping Buffer (Cat. No. 21059)
  • Compatible with many detection systems—western blot, ELISA, and IHC with antibody or avidin/biotin probes (blocker is serum- and biotin-free)
  • Short blocking times—less than 15 minutes for nitrocellulose or PVDF membranes; almost instantaneous for polystyrene microplate wells

Spécifications

Chemical Name or Material PBS
Concentration 1X
For Use With (Application) Western Blot
Physical Form Liquid
Product Line StartingBlock
Quantity 100 mL
Formulation Proprietary protein formulation in phosphate-buffered saline, pH 7.5