Learn More
Brady™ StainerBondz™ Microscope Slide Matte Polyester Labels
Easily create chemical-resistant microscope slide labels with the StainerBondz™ polyester label material (B-481). Print almost anywhere with a Brady™ handheld label printer.
Supplier:  Brady™ M5194481
 Afficher plus de versions de ce produit
Numéro de catalogue. 17012167
Il en reste null
Description
Description
  • Available in presized shapes for commonly used slides and tissue cassettes in which superior chemical resistance is required
  • Withstands the rigors of typical deparaffinization including ethanol and xylene, as well as hematoxylin and eosin slide staining processes
  • Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces in extreme conditions
Spécifications
Identifiants chimiques
Autoclavable
Permanent Acrylic
White
Die-cut
Thermal Transfer
Healthcare and Hospital labelling;General identification;Laboratory labelling
-112°F to 266°F
0.0036 in
0.5 in.
2011/65/EU, 2015/863
None
Satin
Polyester
Non-sterile
-80°C
-80°C to 130°C
0.0914 mm
Spécifications
Autoclavable
Permanent Acrylic
White
Die-cut
Top
Label
Healthcare and Hospital labelling;General identification;Laboratory labelling
-80°C
320 Label(s) / Box
22.86 mm (W) x 19.05 mm (H)
-80°C to 130°C
0.0914 mm
0.75 in.
22.86 mm
0.75 in
19.05 mm
2011/65/EU, 2015/863
None
Satin
Polyester
Thermal Transfer
Non-sterile
BMP51;BMP53;M511;M510
M5-194-481
320
-112°F to 266°F
0.0036 in
22.86 mm (W) x 19.05 mm (H)
0.9 in
22.86 mm
0.5 in.
M5-194-481
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS