missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 0.100 N (N/10), Ricca Chemical
$56.38 - $506.37
Chemical Identifiers
| CAS | 10102-17-7 |
|---|---|
| Molecular Formula | H10Na2O8S2 |
| InChI Key | PODWXQQNRWNDGD-UHFFFAOYSA-L |
| Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
| PubChem CID | 61475 |
| ChEBI | CHEBI:32150 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
7950-4
|
Ricca Chemical Company
79504 |
120 mL | Natural Poly Bottle |
Each for $69.59
|
|
||||
|
795016
|
Ricca Chemical Company
795016 |
500 mL | Natural Poly Bottle |
Each for $56.38
|
|
||||
|
7950-32
|
Ricca Chemical Company
795032 |
1 L | Natural Poly Bottle |
Each for $74.30
|
|
||||
|
7950-1
|
Ricca Chemical Company
79501 |
4 L | Natural Poly Bottle |
Each for $171.75
|
|
||||
|
7950-2-5
|
Ricca Chemical Company
795025 |
10 L | Cubitainer |
Each for $351.84
|
|
||||
|
7950-5
|
Ricca Chemical Company
79505 |
20 L | Cubitainer |
Each for $506.37
|
|
||||
Chemical Identifiers
| 10102-17-7 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| 61475 |
| H10Na2O8S2 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| CHEBI:32150 |
Specifications
| 10102-17-7,7732-18-5,497-19-8 | |
| 1.96,100.0,0.02 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O | |
| 248.17 | |
| CHEBI:32150 | |
| 0.100N | |
| 120 mL |
| 1.88,96.0,0.02 | |
| H10Na2O8S2 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| disodium pentahydrate sulfanidesulfonate | |
| 61475 | |
| ACS/ASTM E 200/AOAC/USP Volumetric | |
| Natural Poly Bottle | |
| Sodium Thiosulfate |