missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 0.100 N (N/10), Ricca Chemical
Supplier: Ricca Chemical Company 79505
Specifications
| Sodium Thiosulfate | |
| 1.88,96.0,0.02 | |
| H10Na2O8S2 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| disodium pentahydrate sulfanidesulfonate | |
| 61475 | |
| ACS/ASTM E 200/AOAC/USP Volumetric | |
| Cubitainer |
| 10102-17-7,7732-18-5,497-19-8 | |
| 1.96,100.0,0.02 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O | |
| 248.17 | |
| CHEBI:32150 | |
| 0.100N | |
| 20 L |
Chemical Identifiers
| 10102-17-7 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| 61475 |
| H10Na2O8S2 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| CHEBI:32150 |