missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.100 N (N/10), Ricca Chemical

Catalog No. 79504
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
79504 120 mL Natural Poly Bottle
795016 500 mL Natural Poly Bottle
795032 1 L Natural Poly Bottle
79501 4 L Natural Poly Bottle
795025 10 L Cubitainer
79505 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 79504 Supplier Ricca Chemical Company Supplier No. 79504

Chemical Identifiers

CAS 10102-17-7 , 7732-18-5 , 497-19-8
Molecular Formula H10Na2O8S2
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150

Specifications

CAS Min % 1.88, 96.0, 0.02
CAS Max % 1.96, 100.0, 0.02
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name disodium pentahydrate sulfanidesulfonate
Molecular Weight (g/mol) 248.17
Grade ACS/ASTM E 200/AOAC/USP Volumetric
Concentration or Composition (by Analyte or Components) 0.100N
Packaging Natural Poly Bottle
Quantity 120 mL
Chemical Name or Material Sodium Thiosulfate