missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 0.100 N (N/10), Ricca Chemical
Specifications
Specifications
| CAS Min % | 1.88, 96.0, 0.02 |
| CAS Max % | 1.96, 100.0, 0.02 |
| SMILES | O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O |
| IUPAC Name | disodium pentahydrate sulfanidesulfonate |
| Molecular Weight (g/mol) | 248.17 |
| Grade | ACS/ASTM E 200/AOAC/USP Volumetric |
| Concentration or Composition (by Analyte or Components) | 0.100N |
| Packaging | Natural Poly Bottle |
| Quantity | 500 mL |
| Chemical Name or Material | Sodium Thiosulfate |