missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate decahydrate, 99+%

CAS: 1303-96-4 | B4Na2O7·10H2O

$62.51 - $568.42

Identifiants chimiques

CAS 1303-96-4
Molecular Formula B4Na2O7·10H2O
MDL Number MFCD00149193
Synonym Borax; Sodium borate (tetra) decahydrate
voir les spécifications

Produits 3
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAA1617636
Afficher les documents
Thermo Scientific Chemicals
A1617636
500 g
chaque for $62.51
Il en reste null
 
AAA161760E
Afficher les documents
Thermo Scientific Chemicals
A161760E
2500 g
chaque for $183.59
Il en reste null
 
AAA161760C
Afficher les documents
Thermo Scientific Chemicals
A161760C
10,000 g
chaque for $568.42
Il en reste null
 
Description

Description

It is used in detergents, cosmetics, and enamel glazes. It is likewise employed to prepare buffer solutions in biochemistry, as a fire retardant, anti-fungal compound, in the manufacture of fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a harbinger for other boron compounds. The molten borax bath is useful for carbide coating process on metals. It is applied as an efficient metal-free catalyst for Hetero-Michael reactions in an aqueous medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in detergents, cosmetics, and enamel glazes. It is likewise employed to prepare buffer solutions in biochemistry, as a fire retardant, anti-fungal compound, in the manufacture of fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a harbinger for other boron compounds. The molten borax bath is useful for carbide coating process on metals. It is applied as an efficient metal-free catalyst for Hetero-Michael reactions in an aqueous medium.

Solubility
Soluble in water, glycerol and ethanol. Insoluble in acids

Notes
Incompatible with strong oxidizing agents and strong acids.
Spécifications

Identifiants chimiques

1303-96-4
B4Na2O7·10H2O
MFCD00149193
Borax; Sodium borate (tetra) decahydrate

Spécifications

75°C
9.24
B4Na2O7·10H2O
MFCD00149193
Borax; Sodium borate (tetra) decahydrate
381.37 (201.22 Anhydrous)
Odorless
Sodium tetraborate decahydrate
1303-96-4
500 g
Na2B4O7·10H2O
14,8590
Soluble in water,glycerol and ethanol. Insoluble in acids
≥99%
1.73
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c

H319-H360FD

missing translation for 'einecsNumber' : 215-540-4

missing translation for 'rtecsNumber' : VZ2275000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only