missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Dibasic, 7-Hydrate, Crystal, U.S.P., J.T. Baker™
$6119.33 - $12391.33
Identifiants chimiques
| CAS | 7782-85-6 |
|---|---|
| Molecular Formula | H15Na2O11P |
| Molecular Weight (g/mol) | 268.06 |
| MDL Number | MFCD00149180 |
| InChI Key | LVXHNCUCBXIIPE-UHFFFAOYSA-L |
| Synonym | sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate |
| PubChem CID | 6096963 |
| IUPAC Name | disodium phosphoric acid heptahydrate |
| SMILES | O.[Na+].[Na+].OP([O-])([O-])=O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
2004208
|
Avantor J.T.Baker
381701 |
500 g | Wide Mouth Amber Glass Bottle |
caisse de 4 chaque for $6,119.33
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
2004206
|
Avantor J.T.Baker
381705 |
2.5 kg | Poly Bottle |
N/A
|
N/A | ||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Identifiants chimiques
| 7782-85-6 | |
| 268.06 | |
| LVXHNCUCBXIIPE-UHFFFAOYSA-L | |
| 6096963 | |
| O.[Na+].[Na+].OP([O-])([O-])=O |
| H15Na2O11P | |
| MFCD00149180 | |
| sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
| disodium phosphoric acid heptahydrate |
Spécifications
| 7782-85-6 | |
| 500 g | |
| MFCD00149180 | |
| LVXHNCUCBXIIPE-UHFFFAOYSA-L | |
| disodium phosphoric acid heptahydrate | |
| 6096963 | |
| USP | |
| 1L = 1.70kg |
| Crystals | |
| H15Na2O11P | |
| sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
| O.[Na+].[Na+].OP([O-])([O-])=O | |
| 268.06 | |
| 268.07 | |
| Wide Mouth Amber Glass Bottle | |
| Sodium Phosphate, Dibasic, 7-Hydrate |