missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, meets analytical spec. of FCC, E251, 99.0-100.5%, Honeywell Fluka™

Numéro de catalogue. p-7071809
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
6001505 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 6001505 Fournisseur Honeywell Inc Code fournisseur 134441KG

Ce produit est controlé donc ne peut être commandé en ligne.
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Meets analytical specification of FCC, E251, 99.0-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Identifiants chimiques

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
PubChem CID 24268
ChEBI CHEBI:63005
SMILES [Na+].[O-][N+]([O-])=O

Spécifications

Quantity 1 kg
Linear Formula NaNO3
UN Number 1498
Solubility Information Soluble in water 1gm/1.1mL, Soluble in alcohol 1gm/125mL, Soluble in boling water 1gm/0.6mL, Soluble in boiling alcohol 1gm/52mL
IUPAC Name sodium;nitrate
Formula Weight 84.99g/mol
Percent Purity 99 to 100.5%
Chemical Name or Material Sodium Nitrate, Meets analytical specification of FCC
Hazard Statement P210-P220-P221-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H319
EINECSNumber 231-554-3
RTECSNumber WC5600000