missing translation for 'onlineSavingsMsg'
Learn More

Sodium Methoxide Solution, MilliporeSigma™ Supelco™

Numéro de catalogue. p-7214143
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111022805 5 mL
111022804 25 mL
111022803 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 111022805 Fournisseur Merck Emd Millipore Code fournisseur 924465ML
Il en reste null

For GC Derivatization

Spécifications

CAS 124-41-4
Quantity 5 mL
Molecular Formula CH3NaO
Linear Formula CH3ONa
MDL Number MFCD00012179
UN Number UN1289 - Class 3 - PG 2 - Sodium Methylate Solutions
Flash Point 9.7°C (Closed Cup)
Synonym Methanolic Base; Sodium Methylate
Formula Weight 54.02
Grade GC Derivatization
Concentration or Composition (by Analyte or Components) 0.5 M
Density 0.800 to 0.812 g/mL (at 20°C)
Afficher plus de résultats Afficher moins de résultats