missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, 0.1M, Honeywell Fluka™

Volumetric, 0.1 M NaOH (0.1N)

$115.32 - $601.37

Identifiants chimiques

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
voir les spécifications

Produits 5
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
6001433
Afficher les documents
Honeywell Inc
319481500ML
500 mL
chaque for $115.32
Il en reste null
 
6001429
Afficher les documents
Honeywell Chemicals
3194811L
1 L
chaque for $158.87
Il en reste null
 
6001432
Afficher les documents
Honeywell Chemicals
3194812L
2 L
chaque for $234.18
Il en reste null
 
6001431
Afficher les documents
Honeywell Inc
31948125L
2.5 L
chaque for $277.79
Il en reste null
 
6001435
Afficher les documents
Honeywell Chemicals
3194816X500ML
6 x 500 mL
caisse de 6 chaque for $601.37
Il en reste null
 
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Spécifications

Identifiants chimiques

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]

Spécifications

1310-73-2
NaOH
0.1M
1g/mL at 25°C
500 mL
[OH-].[Na+]
39.997
CHEBI:32145
>1 (vs air)
Sodium Hydroxide Solution
HNaO
MFCD00003548
NONH for all modes of transport
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40g/mol
3mmHg (37°C)
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'